missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

11-Azidoundecyltrimethoxysilane, 94%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3429803

 View more versions of this product

Catalog No. AAH3429803


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

100°C to 105°C (0.04mmHg)
334521-23-2
C14H31N3O3Si
MFCD11982878
ACSJXAKDVNDMME-UHFFFAOYSA-N
11-azidoundecyl(trimethoxy)silane
46779110
94%
11-Azidoundecyltrimethoxysilane
1 g
N3(CH2)11Si(OCH3)3
11-azidoundecyl trimethoxysilane, 11-trimethoxysilyl undecyl azide
CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC
317.51
317.51
Moisture sensitive

Chemical Identifiers

334521-23-2
317.51
ACSJXAKDVNDMME-UHFFFAOYSA-N
46779110
C14H31N3O3Si
MFCD11982878
11-azidoundecyl trimethoxysilane, 11-trimethoxysilyl undecyl azide
CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only