Mopec™ #11 Sterile Scalpel Blades
High-quality, sterile blades used for anatomic pathology or autopsy.
Supplier:  Mopec™ AJ204
 View more versions of this product
Catalog No. 22-445-602
Explore available promotions
Description
Description
  • Individually wrapped
  • Fits #3, #3L and #5 scalpel handles
Specifications
Specifications
Stainless Steel
# 11
Sterile
No
Scalpel
Single-use
100 Blades/Pk.