missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

12-Azidododecylphosphonic acid, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H33784MD

 View more versions of this product

Catalog No. AAH33784MD


Explore available promotions

Description

Description

This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

Notes
Store in refrigerator. Protect from heat. Incompatible with oxidizing agents.
Specifications

Specifications

12-Azidododecylphosphonic acid
83°C to 85°C
N3(CH2)12P(O)(OH)2
250 mg
C(CCCCCCP(=O)(O)O)CCCCCN=[N+]=[N-]
291.332
291.33
721457-32-5
C12H26N3O3P
MFCD17012690
WHRLXOVKIALVOF-UHFFFAOYSA-N
12-azidododecylphosphonic acid
97638952
95%

Chemical Identifiers

721457-32-5
291.332
WHRLXOVKIALVOF-UHFFFAOYSA-N
12-azidododecylphosphonic acid
C12H26N3O3P
MFCD17012690
97638952
C(CCCCCCP(=O)(O)O)CCCCCN=[N+]=[N-]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only