Learn More
12-Crown-4, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 187020250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chelation/Complexation reagentSpecifications
| 12-Crown-4 | |
| 96.0 | |
| 1.0800g/mL | |
| >112°C | |
| 96% min. (GC) | |
| C8H16O4 | |
| MFCD00005103 | |
| 06,133; 09,126; 12,147; 13,167 | |
| 12-crown-4, eoct, 12-crown-4 ether, ethylene oxide cyclic tetramer, ccris 157, unii-k6069p2c2a, 12-crown4-ether, 12-crown 4-ether, 5-19-11-00334 beilstein handbook reference, 3,6,9,12-tetraoxacyclododecan | |
| XQQZRZQVBFHBHL-UHFFFAOYSA-N | |
| 1,4,7,10-tetraoxacyclododecane | |
| 9269 | |
| 176.21 |
| 294-93-9 | |
| 100.0 | |
| 238°C | |
| Authentic | |
| Glass bottle | |
| 1.4600 to 1.463 | |
| 25g | |
| 1.08 | |
| Solubility in water: miscible | |
| C1COCCOCCOCCO1 | |
| 176.21 | |
| CHEBI:32399 | |
| 97% |
Safety and Handling
GHS H Statement:
Fatal if inhaled.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 206-036-5
RUO â Research Use Only