missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pelican™ 1200 Small Case
Lightweight cas is extremely strong, compact, and fully capable.
$119.48 - $142.29
Specifications
Product Type | Watertight Equipment Case |
---|---|
Material | Polypropylene, Polyurethane Foam |
Depth (English) | 4.87 in. |
Depth (English) Interior | 4.12 in. |
Depth (Metric) | 12.4 cm |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
17-985-414B
|
Pelican™
1200000180 |
Silver |
Each for $119.48
|
|
|||||
17-985-414A
|
Pelican™
1200000110 |
Black |
Each for $142.29
|
|
|||||
Description
- Watertight, crushproof, and dustproof
- Easy open Double Throw latches
- Open cell core with solid wall design makes the case strong yet light weight
- O-ring seal
- Automatic Pressure Equalization Valve balances interior pressure, keeps water out
- Stainless steel hardware
- Pick N Pluck™ with convoluted lid foam
- Lifetime Guarantee of Excellence
Certifications
IP67/TANAG 4280/Def Stan 81-41
Specifications
Watertight Equipment Case | |
4.87 in. | |
12.4 cm | |
10.62 x 9.68 in. (27 x 24.6 cm) | |
10.62 in. | |
27 cm | |
1200 | |
1.2 kg | |
7.12 in. | |
18.1 cm |
Polypropylene, Polyurethane Foam | |
4.12 in. | |
10.5 cm | |
Pick N' Pluck | |
9.25 in. | |
23.5 cm | |
2.45 lb. | |
9.68 in. | |
24.6 cm |