Learn More
Thermo Scientific™ 13mm Autosampler Vial Screw Thread Caps
13mm Screw Thread Caps feature a 13-425 thread finish. Available in an assortment of styles, pre-assembled caps and septa minimize contamination.
$22.02 - $403.23
| Catalog Number | Mfr. No. | Color | Septum | Material | Type | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Septum | Material | Type | Price | Quantity | ||
|
03-375-25A
|
Thermo Scientific™
C40151A |
Black | None | Polypropylene | Open Top Screw Cap |
Pack of 100 Each for $22.02
Case of 10 Pack for $220.20
|
|
||
|
03-378-315
|
Thermo Scientific™
C401566 |
Black | None | Phenolic | Open Top Screw Cap |
Case of 1000 Each for $403.23
|
|
||
|
03-378-316
|
Thermo Scientific™
C401566A |
Black, Red, White | Red PTFE/White Silicone | Phenolic, PTFE, Silicone | Open Top Screw Cap |
Pack of 100 Each for $167.90
|
|
||
|
03-377-106
|
Thermo Scientific™
C401530A |
Black, Ivory, Red | Ivory PTFE/Red Rubber | Polypropylene, PTFE, Rubber | Open Top Screw Cap |
Pack of 100 Each for $118.38
|
|
||
|
03-377-108
|
Thermo Scientific™
C401540A |
Black, Red, White | PTFE/Silicone/PTFE | Polypropylene, PTFE, Silicone | Open Top Screw Cap |
Pack of 100 Each for $170.69
Case of 10 Pack for $1,706.90
|
|
||
|
03-376-250
|
Thermo Scientific™
C401555BLK |
Black, Blue, White | PTFE/Silicone, Pre-slit | Polypropylene, PTFE, Silicone | Open Top Screw Cap |
Pack of 100 Each for $192.69
|
|
||
|
03-375-25C
|
Thermo Scientific™
C40155A |
Natural | Integral Molded | Polypropylene, Polyethylene | Screw Cap |
Pack of 100 Each for $58.38
Case of 10 Pack for $583.80
|
|
||
|
13-622-302
|
Thermo Scientific™
12SCS |
Black | None (no hole) | Polypropylene | Screw Cap |
Pack of 500 Each for $340.70
|
|
||
Description
Thermo Scientific™ 13mm Screw Thread Caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs. Convenient pre-assembled caps and septa minimize contamination from handling. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Autosampler Vial Screw Thread CapsLimited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.