Learn More
Labconco™ 14-Port Manifold; for FreeZone Console Freeze Dryers
Stainless steel manifold for use with FreeZone™ Freeze Dryers includes neoprene valves with plastic knobs.
Supplier:  Labconco™ 7201900
Catalog No. 16-108-561
Only null left
Explore available promotions
Description
Description
14-Port Manifold
  • Type 304 stainless steel manifold
  • 14 neoprene valves with plastic knobs accommodate both 1/2 in. and 3/4 in. adapters
  • For use with 6L, 12L, or 18L FreeZone Console Freeze Dryers
  • Freeze Dryer, glassware, and adapters are required (not included)
  • Includes one latching pin
Specifications
Specifications
Drying Manifold
Stainless Steel (304)
6L, 12L, or 18L FreeZone™ Console Freeze Dryers
24.3 in.
10.6 in.
18 lb.
0
14
14 neoprene valves with molded plastic knobs and 1 latching pin
10.3 in.
21 lb.
Product Suggestions
Product Suggestions