missing translation for 'onlineSavingsMsg'
Learn More
Learn More
missing translation for 'pleaseLogIn'
15-Azido-4,7,10,13-tetraoxapentadecanoic acid, Thermo Scientific™
Chemical Identifiers
CAS | 1257063-35-6 |
---|---|
Molecular Formula | C11H21N3O6 |
Molecular Weight (g/mol) | 291.30 |
MDL Number | MFCD12406155 |
InChI Key | BODPHGOBXPGJKO-UHFFFAOYSA-N |
Synonym | azido-peg4-acid, 15-azido-4,7,10,13-tetraoxapentadecanoic acid, n3-peg 4-cooh, 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid, azido-dpeg r 4-acid, 3-2-2-2-2-azidoethoxy ethoxy ethoxy ethoxy propanoic acid |
PubChem CID | 22731907 |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
AAJ64030MB
|
Thermo Scientific Chemicals
J64030MB |
25mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AAJ64030MC
|
Thermo Scientific Chemicals
J64030MC |
100mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAzido-PEG4-Acid is amine- or phosphine-reactive building block used to derivative primary amines for ligation to alkyne-tagged molecules.
Solubility
Soluble in water.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
1257063-35-6 | |
291.30 | |
BODPHGOBXPGJKO-UHFFFAOYSA-N | |
22731907 |
C11H21N3O6 | |
MFCD12406155 | |
azido-peg4-acid, 15-azido-4,7,10,13-tetraoxapentadecanoic acid, n3-peg 4-cooh, 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid, azido-dpeg r 4-acid, 3-2-2-2-2-azidoethoxy ethoxy ethoxy ethoxy propanoic acid |
Specifications
1257063-35-6 | |
N3CH2CH2(OCH2CH2)4CO2H | |
25mg | |
Soluble in water. | |
291.30 | |
291.3 |
C11H21N3O6 | |
MFCD12406155 | |
azido-peg4-acid, 15-azido-4,7,10,13-tetraoxapentadecanoic acid, n3-peg 4-cooh, 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid, azido-dpeg r 4-acid, 3-2-2-2-2-azidoethoxy ethoxy ethoxy ethoxy propanoic acid | |
BODPHGOBXPGJKO-UHFFFAOYSA-N | |
22731907 | |
15-Azido-4,7,10,13-tetraoxapentadecanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO â Research Use Only