Chemglass Life Sciences 4mL (1 Dram) Vial, 15mm OD x 45mm Height, 13-425 Thread Size
Supplier: Chemglass Inc CG490001
4mL (1 Dram) Vial, 15mm OD x 45mm Height, 13-425 Thread Size. Sample vials packaged in convenient LAB-PACS of 144. Made from Type 1 Borosilicate glass. Full height partitions minimize breakage. Solid PTFE Lined Caps.