Learn More
Encon™ 160 Series Goggles
Impact resistant lens provides distortion-free 160° field of vision
$13.28 - $13.28
Specifications
Certifications/Compliance ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
Lens Tint Clear
Frame Material PVC
View More Specs
Products 1
Catalog Number Mfr. No. Lens Tint Frame Color Includes Price Quantity  
Catalog Number Mfr. No. Lens Tint Frame Color Includes Price Quantity  
19-013960
View Documents
Encon™
05068204
Clear Gray Black Neoprene strap
Each for $13.28
Pack of 12 Each for $159.36
Only null left
Description
Description
  • Clear, polycarbonate cylindrical lens absorbs 99.99% of UV radiation up to 380nm
  • Enfog™ lens coating reduces fogging
  • Large PVC plastic frame accommodates safety spectacles, prescription eyewear, air-purifying respirators, safety caps, and faceshields Indirect ventilation system features vents in top and bottom of frame, with ultrasonic welded top cap; allows airflow while protecting eyes from liquid splash
  • Neoprene strap
Certifications
CSA Z94.3-2007 standards.
Compliance
Meet ANSI Z87.1-2010-High Impact
Specifications
Specifications
ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
Clear
PVC
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'