missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H-1,2,3-Triazolo[4,5-b]pyridine 98.0+%, TCI America™
Supplier: TCI America T13675G
Specifications
1H-1,2,3-Triazolo[4,5-b]pyridine | |
207°C | |
MFCD00134183 | |
4-Azabenzotriazole | |
N1N=C2C=CC=NC2=N1 | |
120.12 | |
120.12 | |
Crystalline Powder |
273-34-7 | |
C5H4N4 | |
5 g | |
VQNDBXJTIJKJPV-UHFFFAOYSA-N | |
2H-[1,2,3]triazolo[4,5-b]pyridine | |
136085 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
273-34-7 | |
120.12 | |
VQNDBXJTIJKJPV-UHFFFAOYSA-N | |
136085 | |
N1N=C2C=CC=NC2=N1 |
C5H4N4 | |
MFCD00134183 | |
4-Azabenzotriazole | |
2H-[1,2,3]triazolo[4,5-b]pyridine |
Safety and Handling
TSCA : No