missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H,1H,2H,2H-Tridecafluoro-n-octyl Acrylate (stabilized with HQ + MEHQ) 98.0+%, TCI America™
$91.06 - $254.30
Chemical Identifiers
CAS | 17527-29-6 |
---|---|
Molecular Formula | C11H7F13O2 |
Molecular Weight (g/mol) | 418.15 |
MDL Number | MFCD00042351 |
InChI Key | VPKQPPJQTZJZDB-UHFFFAOYSA-N |
Synonym | Acrylic Acid 1H,1H,2H,2H-Tridecafluoro-n-octyl Ester |
PubChem CID | 87149 |
IUPAC Name | 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl prop-2-enoate |
SMILES | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C |
Chemical Identifiers
17527-29-6 | |
418.15 | |
VPKQPPJQTZJZDB-UHFFFAOYSA-N | |
87149 | |
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C |
C11H7F13O2 | |
MFCD00042351 | |
Acrylic Acid 1H,1H,2H,2H-Tridecafluoro-n-octyl Ester | |
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl prop-2-enoate |
Specifications
17527-29-6 | |
50°C | |
MFCD00042351 | |
Acrylic Acid 1H,1H,2H,2H-Tridecafluoro-n-octyl Ester | |
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C | |
418.15 | |
418.15 | |
Liquid |
Colorless | |
C11H7F13O2 | |
5 g | |
VPKQPPJQTZJZDB-UHFFFAOYSA-N | |
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl prop-2-enoate | |
87149 | |
≥98.0% (GC) | |
1H,1H,2H,2H-Tridecafluoro-n-octyl Acrylate (stabilized with HQ + MEHQ) |
Safety and Handling
EINECSNumber : (2)-4064
TSCA : Yes