missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,9H-Hexadecafluoro-1-nonanol 97.0+%, TCI America™
Supplier: TCI America H103525G
Specifications
| 1H,1H,9H-Hexadecafluoro-1-nonanol | |
| 59°C | |
| C9H4F16O | |
| 25 g | |
| MSXVQELLSMPBFD-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononan-1-ol | |
| 9779 | |
| ≥97.0% (GC) |
| 376-18-1 | |
| White | |
| MFCD00039629 | |
| 1h,1h,9h-hexadecafluoro-1-nonanol, hexadecafluoro-1-nonanol, 1-nonanol, hexadecafluoro, 1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, 1-nonanol, 1h,1h,9h-hexadecafluoro, 1h,1h,9h-hexadecafluoro-1-hydroxynonane, 1h,1h,9h-perfluoro-1-nonanol, 1h,1h,9h-hexadecafluorononan-1-ol, omega-h-hexadekafluornonanol-1 german, 1h,1h,9h-hexadecafluorononanol | |
| C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
| 432.104 | |
| 432.10 | |
| Crystalline Powder |
Chemical Identifiers
| 376-18-1 | |
| 432.104 | |
| MSXVQELLSMPBFD-UHFFFAOYSA-N | |
| 9779 | |
| C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
| C9H4F16O | |
| MFCD00039629 | |
| 1h,1h,9h-hexadecafluoro-1-nonanol, hexadecafluoro-1-nonanol, 1-nonanol, hexadecafluoro, 1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, 1-nonanol, 1h,1h,9h-hexadecafluoro, 1h,1h,9h-hexadecafluoro-1-hydroxynonane, 1h,1h,9h-perfluoro-1-nonanol, 1h,1h,9h-hexadecafluorononan-1-ol, omega-h-hexadekafluornonanol-1 german, 1h,1h,9h-hexadecafluorononanol | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononan-1-ol |
Safety and Handling
EINECSNumber : (2)-0286
RTECSNumber : RA7905000
TSCA : Yes