Learn More
1H,1H-Perfluorooctylamine, 97%, Thermo Scientific™
$805.52 - $805.52
Chemical Identifiers
CAS | 307-29-9 |
---|---|
Molecular Formula | C8H4F15N |
Molecular Weight (g/mol) | 399.103 |
MDL Number | MFCD00042460 |
InChI Key | HZCZXRSVKNFILZ-UHFFFAOYSA-N |
PubChem CID | 136148 |
IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-amine |
SMILES | C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1684506
|
Thermo Scientific Chemicals
L1684506 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1H,1H-Perfluorooctylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
307-29-9 | |
399.103 | |
HZCZXRSVKNFILZ-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-amine |
C8H4F15N | |
MFCD00042460 | |
136148 | |
C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)N |
Specifications
307-29-9 | |
149°C to 150°C | |
Amine-like | |
1.305 | |
MFCD00042460 | |
UN2735 | |
1h,1h-perfluorooctylamine, 1h,1h-pentadecafluorooctylamine, 1-octanamine, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluorooctylamine, 1h,1h-pentadecafluorooctylamin, 1h,1h-perfluorooctylamine 1g, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-octylamine, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctylamine, 1-octanamine,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro | |
HZCZXRSVKNFILZ-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-amine | |
136148 | |
97% |
1.714 | |
None | |
C8H4F15N | |
CF3(CF2)6CH2NH2 | |
5 g | |
1806966 | |
Not miscible or difficult to mix in water. | |
C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)N | |
399.103 | |
399.1 | |
1H,1H-Perfluorooctylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures