missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole-1-carboxaldehyde 95.0+%, TCI America™

$195.73 - $661.75

Chemical Identifiers

CAS 72773-04-7
Molecular Formula C7H5N3O
Molecular Weight (g/mol) 147.137
MDL Number MFCD00239420
InChI Key DVEIDGKSJOJJJU-UHFFFAOYSA-N
Synonym 1-Formyl-1H-benzotriazole
PubChem CID 576740
IUPAC Name benzotriazole-1-carbaldehyde
SMILES C1=CC=C2C(=C1)N=NN2C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39201G
View Documents
TCI America
B39201G
1 g
Each for $195.73
Only null left
 
B39205G
View Documents
TCI America
B39205G
5 g
Each for $661.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

72773-04-7
147.137
DVEIDGKSJOJJJU-UHFFFAOYSA-N
576740
C1=CC=C2C(=C1)N=NN2C=O
C7H5N3O
MFCD00239420
1-Formyl-1H-benzotriazole
benzotriazole-1-carbaldehyde

Specifications

72773-04-7
White
MFCD00239420
1-Formyl-1H-benzotriazole
C1=CC=C2C(=C1)N=NN2C=O
147.137
147.14
Crystalline Powder
97°C
C7H5N3O
1 g
DVEIDGKSJOJJJU-UHFFFAOYSA-N
benzotriazole-1-carbaldehyde
576740
≥95.0% (GC)
1H-Benzotriazole-1-carboxaldehyde
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'