Learn More
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
$103.33 - $1255.87
Chemical Identifiers
CAS | 95-14-7 |
---|---|
Molecular Formula | C6H5N3 |
Molecular Weight (g/mol) | 119.13 |
MDL Number | MFCD00005699 |
InChI Key | QRUDEWIWKLJBPS-UHFFFAOYSA-N |
Synonym | 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole |
PubChem CID | 7220 |
ChEBI | CHEBI:75331 |
IUPAC Name | 2H-benzotriazole |
SMILES | C1=CC2=NNN=C2C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC183881000
|
Thermo Scientific Chemicals
183881000 |
100 g | Plastic bottle |
Each for $103.33
|
|
||||
AC183885000
|
Thermo Scientific Chemicals
183885000 |
500 g | Plastic bottle |
Each for $262.04
|
|
||||
AC183880025
|
Thermo Scientific Chemicals
183880025 |
2.5 kg | Plastic bottle |
Each for $1,255.87
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
95-14-7 | |
119.13 | |
QRUDEWIWKLJBPS-UHFFFAOYSA-N | |
7220 | |
2H-benzotriazole |
C6H5N3 | |
MFCD00005699 | |
1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole | |
CHEBI:75331 | |
C1=CC2=NNN=C2C=C1 |
Specifications
95-14-7 | |
100.0 | |
Beige-Yellow to White | |
170°C | |
99% | |
C6H5N3 | |
100 g | |
16,21 | |
1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole | |
QRUDEWIWKLJBPS-UHFFFAOYSA-N | |
2H-benzotriazole | |
7220 | |
119.13 | |
Powder, Granules, Crystals, Needles or Flakes |
98.5 | |
94.0°C to 99.0°C | |
204.0°C (15.0 mmHg) | |
Authentic | |
Plastic bottle | |
MFCD00005699 | |
26, 38 | |
15, 1111 | |
Solubility in water: 25g/L in water (20°C). Other solubilities: soluble in alcohol,benzene,toluene,chloroform,and dmf,soluble in ethanol,methanol and ethyleneglycol | |
C1=CC2=NNN=C2C=C1 | |
119.13 | |
CHEBI:75331 | |
99% | |
1H-Benzotriazole, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 202-394-1
RTECSNumber : DM1225000
TSCA : TSCA
RUO – Research Use Only