Learn More
1H-Indazole-4-boronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 430500010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1H-Indazole-4-boronic acid | |
96.0 | |
Authentic | |
Glass Bottle | |
MFCD09878901 | |
1h-indazol-4-yl boronic acid, 1h-indazole-4-boronic acid, indazole-4-boronic acid, 1h-indazol-4-yl-4-boronic acid, 1h-indazol-4-boronic acid, 4-borono-1h-indazole, indazole-4-boronic acid, hcl, indazole-4-boronic acid, hydrochloride, 1h-indazole-4-boronic acid hydrochloride, boronic acid, indazol-4-yl | |
B(C1=C2C=NNC2=CC=C1)(O)O | |
161.96 | |
161.96 |
1023595-17-6 | |
100.0 | |
96% min. (HPLC) | |
C7H7BN2O2 | |
1g | |
BGZZJZIZRARGGZ-UHFFFAOYSA-N | |
1H-indazol-4-ylboronic acid | |
44118310 | |
97% |
Chemical Identifiers
1023595-17-6 | |
161.96 | |
BGZZJZIZRARGGZ-UHFFFAOYSA-N | |
44118310 | |
B(C1=C2C=NNC2=CC=C1)(O)O |
C7H7BN2O2 | |
MFCD09878901 | |
1h-indazol-4-yl boronic acid, 1h-indazole-4-boronic acid, indazole-4-boronic acid, 1h-indazol-4-yl-4-boronic acid, 1h-indazol-4-boronic acid, 4-borono-1h-indazole, indazole-4-boronic acid, hcl, indazole-4-boronic acid, hydrochloride, 1h-indazole-4-boronic acid hydrochloride, boronic acid, indazol-4-yl | |
1H-indazol-4-ylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only