missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H-Indazole-4-carboxaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 669050-70-8 |
---|---|
Molecular Formula | C8H6N2O |
Molecular Weight (g/mol) | 146.15 |
MDL Number | MFCD06738279 |
InChI Key | FPJXNCKSPFGQGC-UHFFFAOYSA-N |
Synonym | 4-formylindazole, 1h-indazole-4-carboxaldehyde, 4-formyl-1h-indazole, 1h-indazol-4-carboxaldehyde, pubchem22512, acmc-209nwx, 1h-indazole-4-carboxaldehyde 9ci |
PubChem CID | 45083569 |
IUPAC Name | 1H-indazole-4-carbaldehyde |
SMILES | C1=CC(=C2C=NNC2=C1)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC431030010
|
Thermo Scientific Chemicals
431030010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC431030050
|
Thermo Scientific Chemicals
431030050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
669050-70-8 | |
146.15 | |
FPJXNCKSPFGQGC-UHFFFAOYSA-N | |
45083569 | |
C1=CC(=C2C=NNC2=C1)C=O |
C8H6N2O | |
MFCD06738279 | |
4-formylindazole, 1h-indazole-4-carboxaldehyde, 4-formyl-1h-indazole, 1h-indazol-4-carboxaldehyde, pubchem22512, acmc-209nwx, 1h-indazole-4-carboxaldehyde 9ci | |
1H-indazole-4-carbaldehyde |
Specifications
669050-70-8 | |
100.0 | |
96% min. (HPLC) | |
C8H6N2O | |
1g | |
FPJXNCKSPFGQGC-UHFFFAOYSA-N | |
1H-indazole-4-carbaldehyde | |
45083569 | |
97% |
96.0 | |
Authentic | |
Glass bottle | |
MFCD06738279 | |
4-formylindazole, 1h-indazole-4-carboxaldehyde, 4-formyl-1h-indazole, 1h-indazol-4-carboxaldehyde, pubchem22512, acmc-209nwx, 1h-indazole-4-carboxaldehyde 9ci | |
C1=CC(=C2C=NNC2=C1)C=O | |
146.15 | |
146.15 | |
1H-Indazole-4-carboxaldehyde |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only