Learn More
1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals
Coupling reagent for preparation of polynucleotides
Supplier: Thermo Scientific Chemicals J61034AO
Description
1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.
Solubility
Miscible with water.
Notes
Incompatible with reducing agents and alkali metals.
Specifications
1H-Tetrazole | |
0.798 | |
MFCD00005247 | |
UN1648 | |
14,2786 | |
Miscible with water. | |
C1=NNN=N1 | |
70.055 | |
CHEBI:33193 | |
0.45M in acetonitrile |
288-94-8 | |
CH2N4 | |
450 mL | |
105799 | |
1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole | |
KJUGUADJHNHALS-UHFFFAOYSA-N | |
2H-tetrazole | |
67519 | |
70.05 | |
Liquid |
Chemical Identifiers
288-94-8 | |
70.055 | |
KJUGUADJHNHALS-UHFFFAOYSA-N | |
CHEBI:33193 | |
C1=NNN=N1 |
CH2N4 | |
MFCD00005247 | |
67519 | |
2H-tetrazole |
Safety and Handling
GHS H Statement
H225-H312-H332-H319
Highly flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
H225-H302+H312+H332-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE
EINECSNumber : 206-023-4
RTECSNumber : UW7370000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only