missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%

CAS: 452304-59-5 | C18H22NP | 283.36 g/mol

$243.62 - $767.90

Chemical Identifiers

CAS 452304-59-5
Molecular Formula C18H22NP
Molecular Weight (g/mol) 283.36
MDL Number MFCD17013996
InChI Key ZATLZEHZPXYMFE-YTXKPMTANA-N
Synonym 1r,2r-2-diphenylphosphino cyclohexanamine, 1r,2r-2-diphenylphosphanyl cyclohexan-1-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 1r,2r-2-diphenylphosphino cyclohexylamine, 1r-2alpha-diphenylphosphino cyclohexane-1beta-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 97+%, 1r,2r-2-diphenylphosphino-1-aminocyclohexane, 1r,2r-2-diphenylphosphino-1-aminocyclohexane,min
PubChem CID 11166387
SMILES N[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60084MD
View Documents
Thermo Scientific Chemicals
H60084MD
250 mg N/A
Only null left
 
AAH6008403
View Documents
Thermo Scientific Chemicals
H6008403
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

452304-59-5
283.36
ZATLZEHZPXYMFE-YTXKPMTANA-N
11166387
C18H22NP
MFCD17013996
1r,2r-2-diphenylphosphino cyclohexanamine, 1r,2r-2-diphenylphosphanyl cyclohexan-1-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 1r,2r-2-diphenylphosphino cyclohexylamine, 1r-2alpha-diphenylphosphino cyclohexane-1beta-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 97+%, 1r,2r-2-diphenylphosphino-1-aminocyclohexane, 1r,2r-2-diphenylphosphino-1-aminocyclohexane,min
N[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

452304-59-5
>110°C (230°F)
MFCD17013996
Air Sensitive
Insoluble in water.
−79° (c=0.5 in Chloroform)
(1R,2R)-2-diphenylphosphanylcyclohexan-1-amine
11166387
≥97%
43°C to 48°C
C18H22NP
250 mg
1r,2r-2-diphenylphosphino cyclohexanamine, 1r,2r-2-diphenylphosphanyl cyclohexan-1-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 1r,2r-2-diphenylphosphino cyclohexylamine, 1r-2alpha-diphenylphosphino cyclohexane-1beta-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 97+%, 1r,2r-2-diphenylphosphino-1-aminocyclohexane, 1r,2r-2-diphenylphosphino-1-aminocyclohexane,min
ZATLZEHZPXYMFE-YTXKPMTANA-N
N[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1
283.36
283.35
(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only