missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol 98.0+%, TCI America™
$130.48 - $699.02
Chemical Identifiers
| CAS | 98919-68-7 |
|---|---|
| Molecular Formula | C12H16O |
| Molecular Weight (g/mol) | 176.259 |
| MDL Number | MFCD00075488 |
| InChI Key | AAIBYZBZXNWTPP-NWDGAFQWSA-N |
| PubChem CID | 102793 |
| IUPAC Name | (1R,2S)-2-phenylcyclohexan-1-ol |
| SMILES | C1CCC(C(C1)C2=CC=CC=C2)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T1490100MG
|
TCI America
T1490100MG |
100 mg |
Each for $130.48
|
|
|||||
|
T14901G
|
TCI America
T14901G |
1 g |
Each for $699.02
|
|
|||||
Chemical Identifiers
| 98919-68-7 | |
| 176.259 | |
| AAIBYZBZXNWTPP-NWDGAFQWSA-N | |
| (1R,2S)-2-phenylcyclohexan-1-ol |
| C12H16O | |
| MFCD00075488 | |
| 102793 | |
| C1CCC(C(C1)C2=CC=CC=C2)O |
Specifications
| 98919-68-7 | |
| White-Yellow | |
| C12H16O | |
| 100 mg | |
| C1CCC(C(C1)C2=CC=CC=C2)O | |
| 176.259 | |
| 176.26 | |
| Crystalline Powder |
| 65°C | |
| 105°C | |
| MFCD00075488 | |
| AAIBYZBZXNWTPP-NWDGAFQWSA-N | |
| (1R,2S)-2-phenylcyclohexan-1-ol | |
| 102793 | |
| ≥98.0% (GC) | |
| (1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol |
Safety and Handling
TSCA : No