missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,3S)-(+)-Camphoric acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2310622

 View more versions of this product

Catalog No. AAB2310622


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(1R,3S)-(+)-Camphoric acid
124-83-4
MFCD00001375
141733
LSPHULWDVZXLIL-LDWIPMOCSA-L
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
6918944
≥98%
+46° (c=1 in Methanol)
183°C to 188°C
C10H14O4-2
2050204
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C
198.218
200.23
100 g

Chemical Identifiers

124-83-4
198.218
LSPHULWDVZXLIL-LDWIPMOCSA-L
6918944
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C
C10H14O4-2
MFCD00001375
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-715-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only