missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol 98.0+%, TCI America™
$98.75 - $298.22
Chemical Identifiers
CAS | 123620-80-4 |
---|---|
Molecular Formula | C13H19NO |
Molecular Weight (g/mol) | 205.30 |
MDL Number | MFCD02093492 |
InChI Key | FZVHJGJBJLFWEX-UHFFFAOYNA-N |
Synonym | 1s,2r-1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl propan-1-ol, 1s,2r-1-phenyl-2-pyrrolidin-1-yl-propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl-1-propanol, 1s,2r-n,n-tetramethylenenorephedrine, 1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r, 1-pyrrolidineethanol, beta-methyl-alpha-phenyl-, alphas,betar, pubchem8276, pubchem14465, 1s,2r-1-phenyl-2-pyrrolizino-1-propanol |
PubChem CID | 9942483 |
IUPAC Name | 1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol |
SMILES | CC(C(O)C1=CC=CC=C1)N1CCCC1 |
Chemical Identifiers
123620-80-4 | |
205.30 | |
FZVHJGJBJLFWEX-UHFFFAOYNA-N | |
9942483 | |
CC(C(O)C1=CC=CC=C1)N1CCCC1 |
C13H19NO | |
MFCD02093492 | |
1s,2r-1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl propan-1-ol, 1s,2r-1-phenyl-2-pyrrolidin-1-yl-propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl-1-propanol, 1s,2r-n,n-tetramethylenenorephedrine, 1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r, 1-pyrrolidineethanol, beta-methyl-alpha-phenyl-, alphas,betar, pubchem8276, pubchem14465, 1s,2r-1-phenyl-2-pyrrolizino-1-propanol | |
1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol |
Specifications
123620-80-4 | |
White-Yellow | |
MFCD02093492 | |
1s,2r-1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl propan-1-ol, 1s,2r-1-phenyl-2-pyrrolidin-1-yl-propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl-1-propanol, 1s,2r-n,n-tetramethylenenorephedrine, 1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r, 1-pyrrolidineethanol, beta-methyl-alpha-phenyl-, alphas,betar, pubchem8276, pubchem14465, 1s,2r-1-phenyl-2-pyrrolizino-1-propanol | |
CC(C(O)C1=CC=CC=C1)N1CCCC1 | |
205.30 | |
205.30 | |
Crystalline Powder |
48°C | |
C13H19NO | |
1 g | |
FZVHJGJBJLFWEX-UHFFFAOYNA-N | |
1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol | |
9942483 | |
≥98.0% (GC) | |
(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol |
Safety and Handling
TSCA : No