missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2595003

 View more versions of this product

Catalog No. AAH2595003


Explore available promotions

Description

Description

(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester is used in organic synthesis as a starting material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester is used in organic synthesis as a starting material and intermediate.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acid and bases.
Specifications

Specifications

(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester
64°C to 67°C
MFCD00757425,MFCD00075490
5262096
Insoluble in water.
COC(=O)C1CC=CCC1C(O)=O
184.19
184.19
88335-93-7
C9H12O4
1 g
1s,2r-1-methyl cis-1,2,3,6-tetrahydrophthalate, 1r,6s-6-methoxycarbonyl cyclohex-3-enecarboxylic acid, 1r,6s-6-methoxycarbonylcyclohex-3-ene-1-carboxylic acid, 1r,6s-6-methoxycarbonyl cyclohex-3-ene-1-carboxylic acid, 1s,2r-1-methyl cis-4-cyclohexene-1,2-dicarboxylate, 1r,6s-6-methoxycarbonyl-1-cyclohex-3-enecarboxylic acid, 1s,2r-1-methylcis-1,2,3,6-tetrahydrophthalate, 2-hydrogen 1-methyl 1s,2r-1,2,3,6-tetrahydrophthalate, 1 r,6s-6-methoxycarbonyl-3-cyclohexene-1-carboxylic acid, 1r, 2s-2-methoxycarbonylcyclohex-4-ene-1-carboxylic acid
MYYLMIDEMAPSGH-UHFFFAOYNA-N
(1R,6S)-6-methoxycarbonylcyclohex-3-ene-1-carboxylic acid
5325907
98%

Chemical Identifiers

88335-93-7
184.19
MYYLMIDEMAPSGH-UHFFFAOYNA-N
5325907
COC(=O)C1CC=CCC1C(O)=O
C9H12O4
MFCD00757425,MFCD00075490
1s,2r-1-methyl cis-1,2,3,6-tetrahydrophthalate, 1r,6s-6-methoxycarbonyl cyclohex-3-enecarboxylic acid, 1r,6s-6-methoxycarbonylcyclohex-3-ene-1-carboxylic acid, 1r,6s-6-methoxycarbonyl cyclohex-3-ene-1-carboxylic acid, 1s,2r-1-methyl cis-4-cyclohexene-1,2-dicarboxylate, 1r,6s-6-methoxycarbonyl-1-cyclohex-3-enecarboxylic acid, 1s,2r-1-methylcis-1,2,3,6-tetrahydrophthalate, 2-hydrogen 1-methyl 1s,2r-1,2,3,6-tetrahydrophthalate, 1 r,6s-6-methoxycarbonyl-3-cyclohexene-1-carboxylic acid, 1r, 2s-2-methoxycarbonylcyclohex-4-ene-1-carboxylic acid
(1R,6S)-6-methoxycarbonylcyclohex-3-ene-1-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only