missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%, Thermo Scientific Chemicals

$112.48 - $1236.33

Chemical Identifiers

CAS 29841-69-8
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00082751
InChI Key PONXTPCRRASWKW-KBPBESRZSA-N
Synonym 1s,2s---1,2-diphenylethylenediamine, 1s,2s---1,2-diphenyl-1,2-ethanediamine, 1s,2s-1,2-diphenylethane-1,2-diamine, 1s,2s-1,2-diphenyl-1,2-ethanediamine, 1s,2s---1,2-diamino-1,2-diphenylethane, s,s-dpen, 1,2-ethanediamine, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethane diamine, --1,2-diphenylethylenediamine, diphenylethylenediamine
PubChem CID 6931238
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diamine
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1296803
View Documents
Thermo Scientific Chemicals
L1296803
1 g
Each for $112.48
 
AAL1296806
View Documents
Thermo Scientific Chemicals
L1296806
5 g
Each for $361.59
 
AAL1296814
View Documents
Thermo Scientific Chemicals
L1296814
25 g
Each for $1,236.33
 
Description

Description

(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air.
Specifications

Chemical Identifiers

29841-69-8
212.296
PONXTPCRRASWKW-KBPBESRZSA-N
6931238
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N
C14H16N2
MFCD00082751
1s,2s---1,2-diphenylethylenediamine, 1s,2s---1,2-diphenyl-1,2-ethanediamine, 1s,2s-1,2-diphenylethane-1,2-diamine, 1s,2s-1,2-diphenyl-1,2-ethanediamine, 1s,2s---1,2-diamino-1,2-diphenylethane, s,s-dpen, 1,2-ethanediamine, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethane diamine, --1,2-diphenylethylenediamine, diphenylethylenediamine
(1S,2S)-1,2-diphenylethane-1,2-diamine

Specifications

29841-69-8
C14H16N2
1 g
3201645
1s,2s---1,2-diphenylethylenediamine, 1s,2s---1,2-diphenyl-1,2-ethanediamine, 1s,2s-1,2-diphenylethane-1,2-diamine, 1s,2s-1,2-diphenyl-1,2-ethanediamine, 1s,2s---1,2-diamino-1,2-diphenylethane, s,s-dpen, 1,2-ethanediamine, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethane diamine, --1,2-diphenylethylenediamine, diphenylethylenediamine
PONXTPCRRASWKW-KBPBESRZSA-N
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N
212.296
212.3
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine
80°C to 84°C
MFCD00082751
UN3259
Air sensitive
Insoluble in water.
−102° (c=1 in Methanol)
(1S,2S)-1,2-diphenylethane-1,2-diamine
6931238
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only