Learn More
(1S,2S)-Cyclohexane-1,2-dicarboxylic acid, 98+%
CAS: 21963-41-7 | C8H12O4 | 172.18 g/mol
$165.95 - $644.34
Chemical Identifiers
| CAS | 21963-41-7 |
|---|---|
| Molecular Formula | C8H12O4 |
| Molecular Weight (g/mol) | 172.18 |
| MDL Number | MFCD00151258,MFCD00001464 |
| InChI Key | QSAWQNUELGIYBC-UHFFFAOYNA-N |
| Synonym | 1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid |
| PubChem CID | 75315 |
| IUPAC Name | (1S,2S)-cyclohexane-1,2-dicarboxylic acid |
| SMILES | OC(=O)C1CCCCC1C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5229003
|
Thermo Scientific Chemicals
H5229003 |
1 g |
Each for $165.95
|
|
|||||
|
AAH5229006
|
Thermo Scientific Chemicals
H5229006 |
5 g |
Each for $644.34
|
|
|||||
Description
A series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.
Solubility
Soluble in acetone(almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 21963-41-7 | |
| 172.18 | |
| QSAWQNUELGIYBC-UHFFFAOYNA-N | |
| 75315 | |
| OC(=O)C1CCCCC1C(O)=O |
| C8H12O4 | |
| MFCD00151258,MFCD00001464 | |
| 1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid | |
| (1S,2S)-cyclohexane-1,2-dicarboxylic acid |
Specifications
| 21963-41-7 | |
| MFCD00151258,MFCD00001464 | |
| 1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid | |
| QSAWQNUELGIYBC-UHFFFAOYNA-N | |
| (1S,2S)-cyclohexane-1,2-dicarboxylic acid | |
| 75315 | |
| ≥98% |
| C8H12O4 | |
| 1 g | |
| Soluble in acetone(almost transparency). | |
| OC(=O)C1CCCCC1C(O)=O | |
| 172.18 | |
| 172.18 | |
| (1S,2S)-Cyclohexane-1,2-dicarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only