missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5274703

 View more versions of this product

Catalog No. AAH5274703


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid
C8H12O4
1 g
FVUHGTQDOMGZOT-NTSWFWBYSA-N
(1S,3R)-3-methoxycarbonylcyclopentane-1-carboxylic acid
10888375
97%
96443-42-4
MFCD04972280
1s,3r-3-methoxycarbonyl cyclopentanecarboxylic acid, 1s,3r-3-methoxycarbonyl cyclopentane-1-carboxylic acid, 1r,3s-1,3-cyclopentanedicarboxylic acid 1-methyl ester, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxylic acid, cis-3-methoxycarbonyl cyclopentancarboxylic acid, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxyl, 1,3-cyclopentanedicarboxylicacid,1-methyl ester, 1r,3s, 1r-cis-1,3-cyclopentanedicarboxylic acid monomethyl ester, 1,3-cyclopentanedicarboxylic acid, 1-methyl ester, 1r,3s
COC(=O)C1CCC(C1)C(=O)O
172.18
172.18

Chemical Identifiers

96443-42-4
172.18
FVUHGTQDOMGZOT-NTSWFWBYSA-N
10888375
COC(=O)C1CCC(C1)C(=O)O
C8H12O4
MFCD04972280
1s,3r-3-methoxycarbonyl cyclopentanecarboxylic acid, 1s,3r-3-methoxycarbonyl cyclopentane-1-carboxylic acid, 1r,3s-1,3-cyclopentanedicarboxylic acid 1-methyl ester, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxylic acid, cis-3-methoxycarbonyl cyclopentancarboxylic acid, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxyl, 1,3-cyclopentanedicarboxylicacid,1-methyl ester, 1r,3s, 1r-cis-1,3-cyclopentanedicarboxylic acid monomethyl ester, 1,3-cyclopentanedicarboxylic acid, 1-methyl ester, 1r,3s
(1S,3R)-3-methoxycarbonylcyclopentane-1-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only