missing translation for 'onlineSavingsMsg'
Learn More

(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one, 95%, 98% ee, Thermo Scientific™

Chemical Identifiers

CAS 134003-03-5
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
InChI Key UIVLZOWDXYXITH-UHNVWZDZSA-N
Synonym 1s,4r-2-azabicyclo 2.2.1 heptan-3-one, 2-azabicyclo 2.2.1 heptan-3-one, 1s,4r, 1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one, 1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
PubChem CID 2734523
IUPAC Name (1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
SMILES C1CC2CC1C(=O)N2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC362222500
SDS View Documents
Thermo Scientific Chemicals
362222500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

134003-03-5
111.14
1s,4r-2-azabicyclo 2.2.1 heptan-3-one, 2-azabicyclo 2.2.1 heptan-3-one, 1s,4r, 1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one, 1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
(1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
C6H9NO
UIVLZOWDXYXITH-UHNVWZDZSA-N
2734523
C1CC2CC1C(=O)N2

Specifications

134003-03-5
Authentic
Glass bottle
250mg
UIVLZOWDXYXITH-UHNVWZDZSA-N
(1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
2734523
95%,98% ee
97.5% min.
94% min. (GC)
C6H9NO
1s,4r-2-azabicyclo 2.2.1 heptan-3-one, 2-azabicyclo 2.2.1 heptan-3-one, 1s,4r, 1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one, 1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
C1CC2CC1C(=O)N2
111.14
111.14
(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one
Videos
SDS
Documents

Documents

RUO – Research Use Only