Learn More
(1S)-(+)-Camphor-10-sulfonic acid, 98+%(dry wt.), water <2%, Thermo Scientific Chemicals
$51.64 - $447.97
Chemical Identifiers
CAS | 3144-16-9 |
---|---|
Molecular Formula | C10H15O4S |
Molecular Weight (g/mol) | 231.29 |
MDL Number | MFCD00064157,MFCD00074827 |
InChI Key | MIOPJNTWMNEORI-XVKPBYJWSA-M |
Synonym | 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid |
PubChem CID | 65617 |
ChEBI | CHEBI:55403 |
IUPAC Name | [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid |
SMILES | CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1617914
|
Thermo Scientific Chemicals
A1617914 |
25 g |
Each for $51.64
|
|
|||||
AAA1617922
|
Thermo Scientific Chemicals
A1617922 |
100 g |
Each for $129.71
|
|
|||||
AAA1617936
|
Thermo Scientific Chemicals
A1617936 |
500 g |
Each for $447.97
|
|
|||||
Description
(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.
Solubility
Exhibit moderate solubility in HCCl.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases. Keep under dry inert gas.
Chemical Identifiers
3144-16-9 | |
231.29 | |
MIOPJNTWMNEORI-XVKPBYJWSA-M | |
65617 | |
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid |
C10H15O4S | |
MFCD00064157,MFCD00074827 | |
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid | |
CHEBI:55403 | |
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2 |
Specifications
3144-16-9 | |
C10H15O4S | |
25 g | |
2809675 | |
14,1734 | |
Exhibit moderate solubility in HCCl. | |
+20° (c=2 in Water) | |
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid | |
65617 | |
232.3 | |
(1S)-(+)-Camphor-10-sulfonic acid |
196°C to 200°C (decomposition) | |
MFCD00064157,MFCD00074827 | |
UN3261 | |
Hygroscopic | |
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid | |
MIOPJNTWMNEORI-XVKPBYJWSA-M | |
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2 | |
231.29 | |
CHEBI:55403 | |
≥98% | |
(dry wt.), water <2% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 221-554-1
RTECSNumber : ED1530000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only