missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150320010

 View more versions of this product

Catalog No. AC150320010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(1S)-(-)-β-Pinene
97.5
-61.0°C
0.8700g/mL
32°C
97.5% min. (GC)
C10H16
MFCD00001345
2319
0.87
− 19.50 (25.00°C c=neat)
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
WTARULDDTDQWMU-IUCAKERBSA-N
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
440967
136.24
Liquid
18172-67-3
100.0
Colorless to Yellow
165.0°C to 167.0°C
Authentic
Glass bottle
1.4770 to 1.4810
1 L
05,154
14,7446
-19.5
Solubility in water: insoluble. Other solubilities: soluble in alcohol,chloroform and ether
CC1(C2CCC(=C)C1C2)C
136.24
CHEBI:28359
98%

Chemical Identifiers

18172-67-3
136.24
WTARULDDTDQWMU-IUCAKERBSA-N
440967
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
C10H16
MFCD00001345
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
CHEBI:28359
CC1(C2CCC(=C)C1C2)C
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
May be fatal if swallowed and enters airways.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed sk

GHS Signal Word: Danger

EINECSNumber : 242-060-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only