Learn More
(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150322500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(1S)-(-)-β-Pinene | |
97.5 | |
-61.0°C | |
0.8700g/mL | |
32°C | |
97.5% min. (GC) | |
C10H16 | |
MFCD00001345 | |
2319 | |
0.87 | |
− 19.50 (25.00°C c=neat) | |
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su | |
WTARULDDTDQWMU-IUCAKERBSA-N | |
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane | |
440967 | |
136.24 | |
Liquid |
18172-67-3 | |
100.0 | |
Colorless to Yellow | |
165.0°C to 167.0°C | |
Authentic | |
Glass bottle | |
1.4770 to 1.4810 | |
250 mL | |
05,154 | |
14,7446 | |
-19.5 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,chloroform and ether | |
CC1(C2CCC(=C)C1C2)C | |
136.24 | |
CHEBI:28359 | |
98% |
Chemical Identifiers
18172-67-3 | |
136.24 | |
WTARULDDTDQWMU-IUCAKERBSA-N | |
440967 | |
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane |
C10H16 | |
MFCD00001345 | |
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su | |
CHEBI:28359 | |
CC1(C2CCC(=C)C1C2)C |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
May be fatal if swallowed and enters airways.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed sk
GHS Signal Word: Danger
EINECSNumber : 242-060-2
RUO – Research Use Only