missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2,1,3-Benzothiadiazole-4-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS
3529-57-5
Molecular Formula
C7H4N2O2S
Molecular Weight (g/mol)
180.181
MDL Number
MFCD05664666
InChI Key
ZGDGZMOKXTUMEV-UHFFFAOYSA-N
Synonym
benzo c 1,2,5 thiadiazole-4-carboxylic acid, 4-carboxybenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole-4-carboxylicacid, 1,2,3-benzothiadiazole-4-carboxylic acid, benzo 1,2,5 thiadiazole-4-carboxylic acid, benzo c 1,2,5-thiadiazole-4-carboxylic acid, 2,1,3-benzothiadiazole-2-siv-4-carboxylic acid, 2??,1,3-benzothiadiazole-4-carboxylic acid
PubChem CID
2795202
IUPAC Name
2,1,3-benzothiadiazole-4-carboxylic acid
SMILES
C1=CC2=NSN=C2C(=C1)C(=O)O
View More Specs
Products
1
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
CC09101DA
SDS
View Documents
Thermo Scientific Chemicals
CC09101DA
1 g
Amber Glass Bottle
Add to cart
Description
Specifications
Chemical Identifiers
CAS
3529-57-5
Molecular Weight (g/mol)
180.181
InChI Key
ZGDGZMOKXTUMEV-UHFFFAOYSA-N
PubChem CID
2795202
SMILES
C1=CC2=NSN=C2C(=C1)C(=O)O
Molecular Formula
C7H4N2O2S
MDL Number
MFCD05664666
Synonym
benzo c 1,2,5 thiadiazole-4-carboxylic acid, 4-carboxybenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole-4-carboxylicacid, 1,2,3-benzothiadiazole-4-carboxylic acid, benzo 1,2,5 thiadiazole-4-carboxylic acid, benzo c 1,2,5-thiadiazole-4-carboxylic acid, 2,1,3-benzothiadiazole-2-siv-4-carboxylic acid, 2??,1,3-benzothiadiazole-4-carboxylic acid
IUPAC Name
2,1,3-benzothiadiazole-4-carboxylic acid
Specifications
CAS
3529-57-5
CAS Max %
100.0
Packaging
Amber Glass Bottle
MDL Number
MFCD05664666
Synonym
benzo c 1,2,5 thiadiazole-4-carboxylic acid, 4-carboxybenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole-4-carboxylicacid, 1,2,3-benzothiadiazole-4-carboxylic acid, benzo 1,2,5 thiadiazole-4-carboxylic acid, benzo c 1,2,5-thiadiazole-4-carboxylic acid, 2,1,3-benzothiadiazole-2-siv-4-carboxylic acid, 2??,1,3-benzothiadiazole-4-carboxylic acid
SMILES
C1=CC2=NSN=C2C(=C1)C(=O)O
Molecular Weight (g/mol)
180.181
Formula Weight
180.19
Chemical Name or Material
2, 1, 3-Benzothiadiazole-4-carboxylic acid
CAS Min %
96.5
Assay Percent Range
97%
Molecular Formula
C7H4N2O2S
Quantity
1 g
InChI Key
ZGDGZMOKXTUMEV-UHFFFAOYSA-N
IUPAC Name
2,1,3-benzothiadiazole-4-carboxylic acid
PubChem CID
2795202
Percent Purity
97%
Videos
Safety and Handling
Safety and Handling
GHS H Statement: May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'