missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,1,3-Benzothiadiazole-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 16405-98-4
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD01647555
InChI Key YHMXJZVGBCACMT-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-5-carboxylic acid, benzo 1,2,5 thiadiazole-5-carboxylic acid, 2??,1,3-benzothiadiazole-5-carboxylic acid, benzo c 1,2,5-thiadiazole-5-carboxylic acid, 4ddm, 2,1,3-benzothiadiazole-5-carboxylicacid, benzo-2,1,3-thiadiazole-5-carboxylic acid, 2,1,3-benzothiadiazole-5-carboxylic acid, benzo 1,2, 5 thiadiazole-5-carboxylic acid, 2$l^ 4 ,1,3-benzothiadiazole-5-carboxylic acid
PubChem CID 602011
IUPAC Name 2,1,3-benzothiadiazole-5-carboxylic acid
SMILES C1=CC2=NSN=C2C=C1C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC09201CB
SDS View Documents
Thermo Scientific Chemicals
CC09201CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC09201DA
SDS View Documents
Thermo Scientific Chemicals
CC09201DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC09201DE
SDS View Documents
Thermo Scientific Chemicals
CC09201DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

16405-98-4
180.181
YHMXJZVGBCACMT-UHFFFAOYSA-N
602011
C1=CC2=NSN=C2C=C1C(=O)O
C7H4N2O2S
MFCD01647555
benzo c 1,2,5 thiadiazole-5-carboxylic acid, benzo 1,2,5 thiadiazole-5-carboxylic acid, 2??,1,3-benzothiadiazole-5-carboxylic acid, benzo c 1,2,5-thiadiazole-5-carboxylic acid, 4ddm, 2,1,3-benzothiadiazole-5-carboxylicacid, benzo-2,1,3-thiadiazole-5-carboxylic acid, 2,1,3-benzothiadiazole-5-carboxylic acid, benzo 1,2, 5 thiadiazole-5-carboxylic acid, 2$l^ 4 ,1,3-benzothiadiazole-5-carboxylic acid
2,1,3-benzothiadiazole-5-carboxylic acid

Specifications

16405-98-4
100.0
Amber Glass Bottle
MFCD01647555
benzo c 1,2,5 thiadiazole-5-carboxylic acid, benzo 1,2,5 thiadiazole-5-carboxylic acid, 2??,1,3-benzothiadiazole-5-carboxylic acid, benzo c 1,2,5-thiadiazole-5-carboxylic acid, 4ddm, 2,1,3-benzothiadiazole-5-carboxylicacid, benzo-2,1,3-thiadiazole-5-carboxylic acid, 2,1,3-benzothiadiazole-5-carboxylic acid, benzo 1,2, 5 thiadiazole-5-carboxylic acid, 2$l^ 4 ,1,3-benzothiadiazole-5-carboxylic acid
C1=CC2=NSN=C2C=C1C(=O)O
180.181
180.19
2, 1, 3-Benzothiadiazole-5-carboxylic acid
96.5
97%
C7H4N2O2S
250mg
YHMXJZVGBCACMT-UHFFFAOYSA-N
2,1,3-benzothiadiazole-5-carboxylic acid
602011
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'