missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,1,3-Benzothiadiazole, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105661000

 View more versions of this product

Catalog No. AC105661000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 1, 3-Benzothiadiazole
97.5
42°C to 44°C
206°C
Authentic
Glass bottle
100 g
piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
C1=CC2=NSN=C2C=C1
136.18
136.18
Low Melting Solid
273-13-2
100.0
Colorless to Yellow
95°C
97.5% min. (GC)
C6H4N2S
27,569
PDQRQJVPEFGVRK-UHFFFAOYSA-N
2,1,3-benzothiadiazole
67502
98%

Chemical Identifiers

273-13-2
136.18
piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
2,1,3-benzothiadiazole
C6H4N2S
PDQRQJVPEFGVRK-UHFFFAOYSA-N
67502
C1=CC2=NSN=C2C=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-985-2

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only