missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroacetamide, 98+%

CAS: 594-65-0 | C2H2Cl3NO | 162.39 g/mol

$125.45 - $125.45

Chemical Identifiers

CAS 594-65-0
Molecular Formula C2H2Cl3NO
Molecular Weight (g/mol) 162.39
MDL Number MFCD00008009
InChI Key UPQQXPKAYZYUKO-UHFFFAOYSA-N
Synonym trichloroacetamide, acetamide, 2,2,2-trichloro, amid kyseliny trichloroctove, acetamide, alpha-trichloro, unii-i8bl3305ro, amid kyseliny trichloroctove czech, 2,2,2-trichloro-acetamide, acetamide, .alpha.-trichloro, .alpha.,.alpha.,.alpha.-trichloroacetamide, trichloro-acetamide
PubChem CID 61144
IUPAC Name 2,2,2-trichloroacetamide
SMILES NC(=O)C(Cl)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC202921000
SDS View Documents
Thermo Scientific Chemicals
202921000
100 g Glass bottle
Each for $125.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

594-65-0
162.39
UPQQXPKAYZYUKO-UHFFFAOYSA-N
61144
NC(=O)C(Cl)(Cl)Cl
C2H2Cl3NO
MFCD00008009
trichloroacetamide, acetamide, 2,2,2-trichloro, amid kyseliny trichloroctove, acetamide, alpha-trichloro, unii-i8bl3305ro, amid kyseliny trichloroctove czech, 2,2,2-trichloro-acetamide, acetamide, .alpha.-trichloro, .alpha.,.alpha.,.alpha.-trichloroacetamide, trichloro-acetamide
2,2,2-trichloroacetamide

Specifications

594-65-0
139°C to 143°C
238°C to 240°C
98% min. (HPLC)
C2H2Cl3NO
MFCD00008009
02, 211
Solubility in water: 13g/L (20°C). Other solubilities: soluble in ethanol,ethylether
NC(=O)C(Cl)(Cl)Cl
162.39
162.4
Powder, Crystals or Chunks
98,98%
White
Authentic
Glass bottle
CCl3CONH2
100 g
trichloroacetamide, acetamide, 2,2,2-trichloro, amid kyseliny trichloroctove, acetamide, alpha-trichloro, unii-i8bl3305ro, amid kyseliny trichloroctove czech, 2,2,2-trichloro-acetamide, acetamide, .alpha.-trichloro, .alpha.,.alpha.,.alpha.-trichloroacetamide, trichloro-acetamide
UPQQXPKAYZYUKO-UHFFFAOYSA-N
2,2,2-trichloroacetamide
61144
98+%
2, 2, 2-Trichloroacetamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-849-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only