Learn More
2,2,2-Trichloroethanol, 99%, Thermo Scientific Chemicals
$65.15 - $233.37
Chemical Identifiers
CAS | 115-20-8 |
---|---|
Molecular Formula | C2H3Cl3O |
Molecular Weight (g/mol) | 149.4 |
MDL Number | MFCD00004677 |
InChI Key | KPWDGTGXUYRARH-UHFFFAOYSA-N |
Synonym | trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol |
PubChem CID | 8259 |
ChEBI | CHEBI:28094 |
IUPAC Name | 2,2,2-trichloroethanol |
SMILES | C(C(Cl)(Cl)Cl)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC139441000
|
Thermo Scientific Chemicals
139441000 |
100 g | Glass bottle |
Each for $65.15
Case of 6 Each for $0.00
|
|
|||||
AC139445000
|
Thermo Scientific Chemicals
139445000 |
500 g | Glass bottle |
Each for $233.37
Case of 6 Each for $0.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
115-20-8 | |
149.4 | |
KPWDGTGXUYRARH-UHFFFAOYSA-N | |
8259 | |
2,2,2-trichloroethanol |
C2H3Cl3O | |
MFCD00004677 | |
trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol | |
CHEBI:28094 | |
C(C(Cl)(Cl)Cl)O |
Specifications
115-20-8 | |
100.0 | |
Colorless to Yellow | |
151.0°C | |
Authentic | |
Glass bottle | |
1.4885 to 1.4905 | |
MFCD00004677 | |
01, 338 | |
1.55 | |
trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol | |
KPWDGTGXUYRARH-UHFFFAOYSA-N | |
2,2,2-trichloroethanol | |
8259 | |
149.4 | |
Liquid |
98.5 | |
17.8°C | |
1.5500g/mL | |
88°C | |
98.5% min. (GC) | |
C2H3Cl3O | |
CCl3CH2OH | |
100 g | |
03,295; 04,521; 06,605 | |
15, 9803 | |
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,ether and carbon,tetrachloride | |
C(C(Cl)(Cl)Cl)O | |
149.4 | |
CHEBI:28094 | |
99% | |
2, 2, 2-Trichloroethanol, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause drowsiness or dizziness.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
Rinse mouth.
IF IN EY
GHS Signal Word: Danger
EINECSNumber : 204-071-0
RTECSNumber : KM3850000
TSCA : TSCA
RUO – Research Use Only