Learn More
2,2,2-Trichloroethyl chloroformate, 97%, Thermo Scientific Chemicals
$96.96 - $264.79
Chemical Identifiers
CAS | 17341-93-4 |
---|---|
Molecular Formula | C3H2Cl4O2 |
Molecular Weight (g/mol) | 211.847 |
MDL Number | MFCD00000810 |
InChI Key | LJCZNYWLQZZIOS-UHFFFAOYSA-N |
Synonym | 2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc |
PubChem CID | 87063 |
IUPAC Name | 2,2,2-trichloroethyl carbonochloridate |
SMILES | C(C(Cl)(Cl)Cl)OC(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0687514
|
Thermo Scientific Chemicals
L0687514 |
25 g |
N/A
|
|
||||||
AAL0687522
|
Thermo Scientific Chemicals
L0687522 |
100 g |
N/A
|
|
||||||
Description
2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.
Solubility
Soluble in Ether, Benzene, Chloroform, Ethanol. Insoluble in water.
Notes
Moisture Sensitive. Desiccate at 4°C. Store under inert gas. Incompatible materials are Strong oxidizing agents.
Chemical Identifiers
17341-93-4 | |
211.847 | |
LJCZNYWLQZZIOS-UHFFFAOYSA-N | |
87063 | |
C(C(Cl)(Cl)Cl)OC(=O)Cl |
C3H2Cl4O2 | |
MFCD00000810 | |
2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc | |
2,2,2-trichloroethyl carbonochloridate |
Specifications
17341-93-4 | |
1.539 | |
>100°C (212°F) | |
1.47 | |
25 g | |
970619 | |
2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc | |
LJCZNYWLQZZIOS-UHFFFAOYSA-N | |
2,2,2-trichloroethyl carbonochloridate | |
87063 | |
97% |
0°C to 2°C | |
171°C to 172°C | |
C3H2Cl4O2 | |
MFCD00000810 | |
UN3277 | |
Moisture sensitive | |
Soluble in Ether,Benzene,Chloroform,Ethanol. Insoluble in water. | |
C(C(Cl)(Cl)Cl)OC(=O)Cl | |
211.847 | |
211.86 | |
2,2,2-Trichloroethyl chloroformate |
Safety and Handling
GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H331-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, N.O.S.
EINECSNumber : 241-363-7
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only