missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoro-3'-nitroacetophenone 98.0+%, TCI America™
$175.22 - $521.94
Chemical Identifiers
| CAS | 657-15-8 |
|---|---|
| Molecular Formula | C8H4F3NO3 |
| Molecular Weight (g/mol) | 219.119 |
| MDL Number | MFCD00034541 |
| InChI Key | QADCNGZPRUSTJL-UHFFFAOYSA-N |
| PubChem CID | 69566 |
| IUPAC Name | 2,2,2-trifluoro-1-(3-nitrophenyl)ethanone |
| SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)C(F)(F)F |
Chemical Identifiers
| 657-15-8 | |
| 219.119 | |
| QADCNGZPRUSTJL-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-(3-nitrophenyl)ethanone |
| C8H4F3NO3 | |
| MFCD00034541 | |
| 69566 | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)C(F)(F)F |
Specifications
| 657-15-8 | |
| 113°C | |
| MFCD00034541 | |
| QADCNGZPRUSTJL-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-(3-nitrophenyl)ethanone | |
| 69566 | |
| ≥98.0% (GC) | |
| 2,2,2-Trifluoro-3′-nitroacetophenone |
| 56°C | |
| C8H4F3NO3 | |
| 1 g | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)C(F)(F)F | |
| 219.119 | |
| 219.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No