missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2,2-Trifluoro-N,N-dimethylacetamide 98.0+%, TCI America™
$82.44 - $324.76
Chemical Identifiers
CAS | 1547-87-1 |
---|---|
Molecular Formula | C4H6F3NO |
Molecular Weight (g/mol) | 141.09 |
MDL Number | MFCD00043555 |
InChI Key | WXBWKMLIVXELSF-UHFFFAOYSA-N |
PubChem CID | 350491 |
IUPAC Name | 2,2,2-trifluoro-N,N-dimethylacetamide |
SMILES | CN(C)C(=O)C(F)(F)F |
Chemical Identifiers
1547-87-1 | |
141.09 | |
WXBWKMLIVXELSF-UHFFFAOYSA-N | |
2,2,2-trifluoro-N,N-dimethylacetamide |
C4H6F3NO | |
MFCD00043555 | |
350491 | |
CN(C)C(=O)C(F)(F)F |
Specifications
1547-87-1 | |
136°C | |
MFCD00043555 | |
1993 | |
CN(C)C(=O)C(F)(F)F | |
141.09 | |
141.09 | |
Liquid |
Colorless | |
C4H6F3NO | |
5 g | |
WXBWKMLIVXELSF-UHFFFAOYSA-N | |
2,2,2-trifluoro-N,N-dimethylacetamide | |
350491 | |
≥98.0% (GC) | |
2,2,2-Trifluoro-N,N-dimethylacetamide |
Safety and Handling
TSCA : No