missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanesulfonyl chloride, 95%

CAS: 1648-99-3 | C2H2ClF3O2S | 182.55 g/mol

$189.74 - $492.99

Chemical Identifiers

CAS 1648-99-3
Molecular Formula C2H2ClF3O2S
Molecular Weight (g/mol) 182.55
MDL Number MFCD00007458
InChI Key CXCHEKCRJQRVNG-UHFFFAOYSA-N
Synonym tresyl chloride, 2,2,2-trifluoroethane-1-sulfonyl chloride, ethanesulfonyl chloride, 2,2,2-trifluoro, 2,2,2-trifluoroethanesulphonyl chloride, 2,2,2-trifluoroethyl sulphonyl chloride, 2,2,2-trifluoroethylsulfonyl chloride, chloro 2,2,2-trifluoroethyl sulfone, 2,2,2-trifluoroethanesulfonylchloride, ethanesulfonylchloride, 2,2,2-trifluoro, pubchem2553
PubChem CID 74242
IUPAC Name 2,2,2-trifluoroethanesulfonyl chloride
SMILES C(C(F)(F)F)S(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC305350010
SDS View Documents
Thermo Scientific Chemicals
305350010
1 g Glass bottle
Each for $189.74
Only null left
 
AC305350025
SDS View Documents
Thermo Scientific Chemicals
305350025
2.5 g Glass bottle
Each for $492.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1648-99-3
182.55
CXCHEKCRJQRVNG-UHFFFAOYSA-N
74242
C(C(F)(F)F)S(=O)(=O)Cl
C2H2ClF3O2S
MFCD00007458
tresyl chloride, 2,2,2-trifluoroethane-1-sulfonyl chloride, ethanesulfonyl chloride, 2,2,2-trifluoro, 2,2,2-trifluoroethanesulphonyl chloride, 2,2,2-trifluoroethyl sulphonyl chloride, 2,2,2-trifluoroethylsulfonyl chloride, chloro 2,2,2-trifluoroethyl sulfone, 2,2,2-trifluoroethanesulfonylchloride, ethanesulfonylchloride, 2,2,2-trifluoro, pubchem2553
2,2,2-trifluoroethanesulfonyl chloride

Specifications

1648-99-3
100.0
1.6500g/mL
72°C
98.5% min. (ex Cl) (Argentometry)
C2H2ClF3O2S
CF3CH2SO2Cl
1 g
tresyl chloride, 2,2,2-trifluoroethane-1-sulfonyl chloride, ethanesulfonyl chloride, 2,2,2-trifluoro, 2,2,2-trifluoroethanesulphonyl chloride, 2,2,2-trifluoroethyl sulphonyl chloride, 2,2,2-trifluoroethylsulfonyl chloride, chloro 2,2,2-trifluoroethyl sulfone, 2,2,2-trifluoroethanesulfonylchloride, ethanesulfonylchloride, 2,2,2-trifluoro, pubchem2553
C(C(F)(F)F)S(=O)(=O)Cl
182.55
182.55
Liquid
98.5
Colorless
64°C to 67°C (45.0 mmHg)
Authentic
Glass bottle
1.3870 to 1.389
MFCD00007458
1.65
CXCHEKCRJQRVNG-UHFFFAOYSA-N
2,2,2-trifluoroethanesulfonyl chloride
74242
99%
2, 2, 2-Trifluoroethanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-713-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only