missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2,2-Trifluoroethyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
Supplier: TCI America A11525G
Specifications
2,2,2-Trifluoroethyl Acrylate (stabilized with MEHQ) | |
Colorless | |
C5H5F3O2 | |
5 g | |
2,2,2-trifluoroethyl acrylate, 2-propenoic acid, 2,2,2-trifluoroethyl ester, 2-propenoic acid, trifluoroethyl ester, 2,2,2-trifluoroethylacrylate, acrylic acid, 2,2,2-trifluoroethyl ester, acrylic acid 2,2,2-trifluoroethyl ester, tfol-a, acmc-20aokw, trifluoroethyl acrylate, pubchem12647 | |
C=CC(=O)OCC(F)(F)F | |
154.088 | |
154.09 | |
Liquid |
407-47-6 | |
92°C | |
MFCD00000444 | |
3272 | |
VBHXIMACZBQHPX-UHFFFAOYSA-N | |
2,2,2-trifluoroethyl prop-2-enoate | |
67889 | |
≥98.0% (GC) |
Chemical Identifiers
407-47-6 | |
154.088 | |
VBHXIMACZBQHPX-UHFFFAOYSA-N | |
67889 | |
C=CC(=O)OCC(F)(F)F |
C5H5F3O2 | |
MFCD00000444 | |
2,2,2-trifluoroethyl acrylate, 2-propenoic acid, 2,2,2-trifluoroethyl ester, 2-propenoic acid, trifluoroethyl ester, 2,2,2-trifluoroethylacrylate, acrylic acid, 2,2,2-trifluoroethyl ester, acrylic acid 2,2,2-trifluoroethyl ester, tfol-a, acmc-20aokw, trifluoroethyl acrylate, pubchem12647 | |
2,2,2-trifluoroethyl prop-2-enoate |
Safety and Handling
TSCA : Yes