Learn More
2,2,2-Trifluoroethylamine, 99.5%
CAS: 753-90-2 | C2H4F3N | 99.06 g/mol
$94.80 - $614.05
Chemical Identifiers
| CAS | 753-90-2 |
|---|---|
| Molecular Formula | C2H4F3N |
| Molecular Weight (g/mol) | 99.06 |
| MDL Number | MFCD00008132 |
| InChI Key | KIPSRYDSZQRPEA-UHFFFAOYSA-N |
| Synonym | 2,2,2-trifluoroethylamine, trifluoroethylamine, ethanamine, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-amine, 2-amino-1,1,1-trifluoroethane, ethylamine, 2,2,2-trifluoro, 2,2,2-trifluoro-ethylamine, 2,2,2-trifluorethanamin, trifluoro ethylamine, trifluoroethyl amine |
| PubChem CID | 9773 |
| IUPAC Name | 2,2,2-trifluoroethanamine |
| SMILES | C(C(F)(F)F)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC303500010
|
Thermo Scientific Chemicals
303500010 |
1 g | Glass bottle |
Each for $94.80
|
|
||||
|
AC303500100
|
Thermo Scientific Chemicals
303500100 |
10 g | Glass bottle |
Each for $614.05
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 753-90-2 | |
| 99.06 | |
| KIPSRYDSZQRPEA-UHFFFAOYSA-N | |
| 9773 | |
| C(C(F)(F)F)N |
| C2H4F3N | |
| MFCD00008132 | |
| 2,2,2-trifluoroethylamine, trifluoroethylamine, ethanamine, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-amine, 2-amino-1,1,1-trifluoroethane, ethylamine, 2,2,2-trifluoro, 2,2,2-trifluoro-ethylamine, 2,2,2-trifluorethanamin, trifluoro ethylamine, trifluoroethyl amine | |
| 2,2,2-trifluoroethanamine |
Specifications
| 753-90-2 | |
| 100.0 | |
| 1.2400g/mL | |
| -16°C | |
| 99.4% min. (GC) | |
| C2H4F3N | |
| CF3CH2NH2 | |
| 1 g | |
| 2,2,2-trifluoroethylamine, trifluoroethylamine, ethanamine, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-amine, 2-amino-1,1,1-trifluoroethane, ethylamine, 2,2,2-trifluoro, 2,2,2-trifluoro-ethylamine, 2,2,2-trifluorethanamin, trifluoro ethylamine, trifluoroethyl amine | |
| C(C(F)(F)F)N | |
| 99.06 | |
| 99.06 | |
| Liquid |
| 99.4 | |
| Colorless | |
| 37°C to 38°C | |
| Authentic | |
| Glass bottle | |
| 1.3000 to 1.302 | |
| MFCD00008132 | |
| 1.24 | |
| KIPSRYDSZQRPEA-UHFFFAOYSA-N | |
| 2,2,2-trifluoroethanamine | |
| 9773 | |
| 99.50% | |
| 2, 2, 2-Trifluoroethylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Highly flammable liquid and vapor.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f
GHS Signal Word: Danger
EINECSNumber : 212-041-3
RUO – Research Use Only