missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,3,3,4,4-Hexafluoro-1,5-pentanediol 98.0+%, TCI America™

Supplier:  TCI America H074625G

 View more versions of this product

Catalog No. H074625G


Explore available promotions

Description

Description

Specifications

Specifications

2,2,3,3,4,4-Hexafluoro-1,5-pentanediol
79°C
112°C
MFCD00042434
2,2,3,3,4,4-hexafluoro-1,5-pentanediol, hexafluoroamylene glycol, 1,5-pentanediol, 2,2,3,3,4,4-hexafluoro, hexafluoro-1,5-pentanediol, 2,2,3,3,4,4-hexafluoropentanediol, 2,2,3,3,4,4-hexafluoropentan-1,5-diol, 1,5-dihydroxy-2,2,3,3,4,4-hexafluoropentane, acmc-209iuy, ielvmupswdzwsd-uhfffaoysa, 2,3,3,4,4-hexafluoropentanediol
C(C(C(C(CO)(F)F)(F)F)(F)F)O
212.091
212.09
Crystalline Powder
376-90-9
White
C5H6F6O2
25 g
IELVMUPSWDZWSD-UHFFFAOYSA-N
2,2,3,3,4,4-hexafluoropentane-1,5-diol
67831
≥98.0% (GC)

Chemical Identifiers

376-90-9
212.091
IELVMUPSWDZWSD-UHFFFAOYSA-N
67831
C(C(C(C(CO)(F)F)(F)F)(F)F)O
C5H6F6O2
MFCD00042434
2,2,3,3,4,4-hexafluoro-1,5-pentanediol, hexafluoroamylene glycol, 1,5-pentanediol, 2,2,3,3,4,4-hexafluoro, hexafluoro-1,5-pentanediol, 2,2,3,3,4,4-hexafluoropentanediol, 2,2,3,3,4,4-hexafluoropentan-1,5-diol, 1,5-dihydroxy-2,2,3,3,4,4-hexafluoropentane, acmc-209iuy, ielvmupswdzwsd-uhfffaoysa, 2,3,3,4,4-hexafluoropentanediol
2,2,3,3,4,4-hexafluoropentane-1,5-diol
Videos
Safety and Handling

Safety and Handling

RTECSNumber : SA0750000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions