Learn More
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D, Thermo Scientific Chemicals
$146.25 - $1291.45
Chemical Identifiers
CAS | 24493-21-8 |
---|---|
Molecular Formula | C6H13NaO2Si |
Molecular Weight (g/mol) | 172.267 |
MDL Number | MFCD00002762 |
InChI Key | OIIWPAYIXDCDNL-HGFPCDIYSA-M |
Synonym | sodium 3-trimethylsilyl 2,2,3,3-2h4 propionate, 3-trimethylsilyl propionic acid-d4 sodium salt, 2,2,3,3-d4-3-trimethylsilyl propionic acid sodium salt, sodium 3-trimethylsilyl 2 h? propanoate, sodium 3-trimethylsilyl 2h4 propanoate, tsp-d4, propanoic-2,2,3,3-d4 acid, 3-trimethylsilyl-, sodium salt, sodium 3-trimethylsilyl propionate-2,2,3,3-d4, 3-trimethylsilyl propionic-2,2,3,3-d4 acid sodium salt, sodium-3-trimethylsilyl-2,2,3,3-tetradeuteriopropionate |
PubChem CID | 23688921 |
IUPAC Name | sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate |
SMILES | C[Si](C)(C)CCC(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA14489MD
|
Thermo Scientific Chemicals
A14489MD |
250 mg |
Each for $146.25
|
|
|||||
AAA1448903
|
Thermo Scientific Chemicals
A1448903 |
1 g |
Each for $285.25
|
|
|||||
AAA1448906
|
Thermo Scientific Chemicals
A1448906 |
5 g |
Each for $1,291.45
|
|
|||||
Description
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.
Solubility
Soluble in water.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
Chemical Identifiers
24493-21-8 | |
172.267 | |
OIIWPAYIXDCDNL-HGFPCDIYSA-M | |
23688921 | |
C[Si](C)(C)CCC(=O)[O-].[Na+] |
C6H13NaO2Si | |
MFCD00002762 | |
sodium 3-trimethylsilyl 2,2,3,3-2h4 propionate, 3-trimethylsilyl propionic acid-d4 sodium salt, 2,2,3,3-d4-3-trimethylsilyl propionic acid sodium salt, sodium 3-trimethylsilyl 2 h? propanoate, sodium 3-trimethylsilyl 2h4 propanoate, tsp-d4, propanoic-2,2,3,3-d4 acid, 3-trimethylsilyl-, sodium salt, sodium 3-trimethylsilyl propionate-2,2,3,3-d4, 3-trimethylsilyl propionic-2,2,3,3-d4 acid sodium salt, sodium-3-trimethylsilyl-2,2,3,3-tetradeuteriopropionate | |
sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate |
Specifications
24493-21-8 | |
C6H13NaO2Si | |
MFCD00002762 | |
4046239 | |
sodium 3-trimethylsilyl 2,2,3,3-2h4 propionate, 3-trimethylsilyl propionic acid-d4 sodium salt, 2,2,3,3-d4-3-trimethylsilyl propionic acid sodium salt, sodium 3-trimethylsilyl 2 h? propanoate, sodium 3-trimethylsilyl 2h4 propanoate, tsp-d4, propanoic-2,2,3,3-d4 acid, 3-trimethylsilyl-, sodium salt, sodium 3-trimethylsilyl propionate-2,2,3,3-d4, 3-trimethylsilyl propionic-2,2,3,3-d4 acid sodium salt, sodium-3-trimethylsilyl-2,2,3,3-tetradeuteriopropionate | |
OIIWPAYIXDCDNL-HGFPCDIYSA-M | |
sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate | |
23688921 | |
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt |
>300°C | |
(CH3)3SiCD2CD2CO2Na | |
250 mg | |
Hygroscopic | |
Soluble in water. | |
C[Si](C)(C)CCC(=O)[O-].[Na+] | |
172.267 | |
172.27 | |
≥98 atom %D |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-286-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only