Learn More
2,2,3,3-Tetrafluoro-1-propanol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2159418
Description
2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications
2,2,3,3-Tetrafluoro-1-propanol | |
<-70°C | |
107°C to 109°C | |
C3H4F4O | |
MFCD00004676 | |
UN1987 | |
14,9206 | |
Soluble in water. | |
OCC(F)(F)C(F)F | |
132.06 | |
132.06 |
76-37-9 | |
1.471 | |
49°C (120°F) | |
1.3195 | |
50 g | |
1738264 | |
2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff | |
NBUKAOOFKZFCGD-UHFFFAOYSA-N | |
2,2,3,3-tetrafluoropropan-1-ol | |
6441 | |
97% |
Chemical Identifiers
76-37-9 | |
132.06 | |
NBUKAOOFKZFCGD-UHFFFAOYSA-N | |
6441 | |
OCC(F)(F)C(F)F |
C3H4F4O | |
MFCD00004676 | |
2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff | |
2,2,3,3-tetrafluoropropan-1-ol |
Safety and Handling
GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 200-955-5
RTECSNumber : UB9400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only