missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2',3,3'-Tetrahydroxy-1,1'-binaphthyl 98.0+%, TCI America™

Supplier:  TCI America T17541G

Catalog No. T17541G


Explore available promotions

Description

Description

Specifications

Specifications

2,2′,3,3′-Tetrahydroxy-1,1′-binaphthyl
Red-White
MFCD00188003
1,1′C-Bi(2,3-naphthodiol)
OC1=C(O)C(=C2C=CC=CC2=C1)C1=C2C=CC=CC2=CC(O)=C1O
318.33
318.33
Crystalline Powder
39215-21-9
C20H14O4
1 g
OOSHJGKXQBJASF-UHFFFAOYSA-N
[1,1'-binaphthalene]-2,2',3,3'-tetrol
316673
≥98.0% (HPLC)

Chemical Identifiers

39215-21-9
318.33
OOSHJGKXQBJASF-UHFFFAOYSA-N
316673
OC1=C(O)C(=C2C=CC=CC2=C1)C1=C2C=CC=CC2=CC(O)=C1O
C20H14O4
MFCD00188003
1,1′C-Bi(2,3-naphthodiol)
[1,1'-binaphthalene]-2,2',3,3'-tetrol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions