missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(2,4-Dichlorophenoxy)propionic Acid 98.0+%, TCI America™
Supplier: TCI America D194225G
Specifications
2-(2,4-Dichlorophenoxy)propionic Acid | |
115°C | |
MFCD00002645 | |
3077 | |
MZHCENGPTKEIGP-UHFFFAOYNA-N | |
2-(2,4-dichlorophenoxy)propanoic acid | |
8427 | |
235.06 | |
Crystalline Powder |
120-36-5 | |
C9H8Cl2O3 | |
25 g | |
dichlorprop, 2-2,4-dichlorophenoxy propanoic acid, 2-2,4-dichlorophenoxy propionic acid, hormatox, polymone, dichloroprop, polytox, kildip, celatox-dp, herbizid dp | |
CC(OC1=CC=C(Cl)C=C1Cl)C(O)=O | |
235.06 | |
CHEBI:75370 | |
≥98.0% (T) |
Chemical Identifiers
120-36-5 | |
235.06 | |
MZHCENGPTKEIGP-UHFFFAOYNA-N | |
8427 | |
2-(2,4-dichlorophenoxy)propanoic acid |
C9H8Cl2O3 | |
MFCD00002645 | |
dichlorprop, 2-2,4-dichlorophenoxy propanoic acid, 2-2,4-dichlorophenoxy propionic acid, hormatox, polymone, dichloroprop, polytox, kildip, celatox-dp, herbizid dp | |
CHEBI:75370 | |
CC(OC1=CC=C(Cl)C=C1Cl)C(O)=O |
Safety and Handling
RTECSNumber : UF1050000
TSCA : Yes