missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol Diisobutyrate 98.5+%, TCI America™

$29.83 - $42.87

Chemical Identifiers

CAS 6846-50-0
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.412
MDL Number MFCD00059267
InChI Key OMVSWZDEEGIJJI-UHFFFAOYSA-N
Synonym 2,2,4-trimethyl-1,3-pentanediol diisobutyrate, kodaflex txib, 2,2,4-trimethylpentane-1,3-diyl bis 2-methylpropanoate, txib, propanoic acid, 2-methyl-, 2,2-dimethyl-1-1-methylethyl-1,3-propanediyl ester, 2,2,4-trimethylpentanediol-1,3-diisobutyrate, isobutyric acid, 1-isopropyl-2,2-dimethyltrimethylene ester, 1-isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-trimethyl-1,3-pentanediol ester, dsstox_cid_7635
PubChem CID 23284
IUPAC Name [2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate
SMILES CC(C)C(C(C)(C)COC(=O)C(C)C)OC(=O)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T099725ML
View Documents
TCI America
T099725ML
25 mL
Each for $29.83
Only null left
 
T0997500ML
View Documents
TCI America
T0997500ML
500 mL
Each for $42.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6846-50-0
286.412
OMVSWZDEEGIJJI-UHFFFAOYSA-N
23284
CC(C)C(C(C)(C)COC(=O)C(C)C)OC(=O)C(C)C
C16H30O4
MFCD00059267
2,2,4-trimethyl-1,3-pentanediol diisobutyrate, kodaflex txib, 2,2,4-trimethylpentane-1,3-diyl bis 2-methylpropanoate, txib, propanoic acid, 2-methyl-, 2,2-dimethyl-1-1-methylethyl-1,3-propanediyl ester, 2,2,4-trimethylpentanediol-1,3-diisobutyrate, isobutyric acid, 1-isopropyl-2,2-dimethyltrimethylene ester, 1-isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-trimethyl-1,3-pentanediol ester, dsstox_cid_7635
[2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate

Specifications

6846-50-0
Colorless
C16H30O4
25 mL
OMVSWZDEEGIJJI-UHFFFAOYSA-N
[2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate
23284
≥98.5% (GC)
2,2,4-Trimethyl-1,3-pentanediol Diisobutyrate
-70°C
280°C
MFCD00059267
2,2,4-trimethyl-1,3-pentanediol diisobutyrate, kodaflex txib, 2,2,4-trimethylpentane-1,3-diyl bis 2-methylpropanoate, txib, propanoic acid, 2-methyl-, 2,2-dimethyl-1-1-methylethyl-1,3-propanediyl ester, 2,2,4-trimethylpentanediol-1,3-diisobutyrate, isobutyric acid, 1-isopropyl-2,2-dimethyltrimethylene ester, 1-isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-trimethyl-1,3-pentanediol ester, dsstox_cid_7635
CC(C)C(C(C)(C)COC(=O)C(C)C)OC(=O)C(C)C
286.412
286.41
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2498

RTECSNumber : SA1420000

TSCA : Yes

SDS
Documents

Documents