missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ACS, 99+%

CAS: 540-84-1 | C8H18 | 114.232 g/mol

$85.26 - $374.25

Identifiants chimiques

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C
voir les spécifications

Produits 4
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA31787AP
Afficher les documents
Thermo Scientific Chemicals
031787AP
500 mL
chaque for $85.26
Il en reste null
 
AA31787K2
Afficher les documents
Thermo Scientific Chemicals
031787K2
1 L
chaque for $119.68
Il en reste null
 
AA31787M1
Afficher les documents
Thermo Scientific Chemicals
031787M1
4 x 1 L
chaque for $374.25
Il en reste null
 
AA31787K7
Afficher les documents
Thermo Scientific Chemicals
031787K7
4 L
chaque for $268.33
Il en reste null
 
Description

Description

Solvent, thinner, in determination of fuel octane numbers2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as a thinner. It is involved as a processing aid in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

540-84-1
114.232
NHTMVDHEPJAVLT-UHFFFAOYSA-N
10907
2,2,4-trimethylpentane
C8H18
MFCD00008943
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CHEBI:62805
CC(C)CC(C)(C)C

Spécifications

540-84-1
Colorless
98°C to 99°C
>99%
C8H18
MFCD00008943
1262
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
NHTMVDHEPJAVLT-UHFFFAOYSA-N
2,2,4-trimethylpentane
10907
114.23
ACS
2, 2, 4-Trimethylpentane
-107°C
0.692
-7°C (19°F)
Nearly Odorless
1.391
500 mL
1696876
Soluble in benzene,toluene,xylene,chloroform,ether,CS2,CCl4,and DMF. Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,acetone,heptane and oils. Immiscible with water.
CC(C)CC(C)(C)C
114.232
CHEBI:62805
15 hPa (11mm Hg) at 20°C
Liquid
Videos
Sécurité et manipulation

Sécurité et manipulation

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336

missing translation for 'dotInformation' : DOT Class: 3; Packing Group: II

missing translation for 'einecsNumber' : 208-759-1

missing translation for 'rtecsNumber' : SA3320000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only