Learn More
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%, Thermo Scientific Chemicals
$151.10 - $1193.11
Chemical Identifiers
CAS | 540-84-1 |
---|---|
Molecular Formula | C8H18 |
Molecular Weight (g/mol) | 114.232 |
MDL Number | MFCD00008943 |
InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
Synonym | isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane |
PubChem CID | 10907 |
ChEBI | CHEBI:62805 |
IUPAC Name | 2,2,4-trimethylpentane |
SMILES | CC(C)CC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA22901K2
|
Thermo Scientific Chemicals
022901K2 |
1 L |
Each for $151.10
|
|
|||||
AA22901M1
|
Thermo Scientific Chemicals
22901M1 |
4 x 1 L |
Each for $434.63
|
|
|||||
AA22901K7
|
Thermo Scientific Chemicals
022901K7 |
4 L |
Each for $392.01
|
|
|||||
AA22901M6
|
Thermo Scientific Chemicals
022901M6 |
4 x 4 L |
Each for $1,193.11
|
|
|||||
Description
2,2,4-Trimethylpentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
540-84-1 | |
114.232 | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
10907 | |
2,2,4-trimethylpentane |
C8H18 | |
MFCD00008943 | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
CHEBI:62805 | |
CC(C)CC(C)(C)C |
Specifications
540-84-1 | |
Colorless | |
98°C to 99°C | |
410°C (770°F) | |
Nearly odorless | |
1.391 | |
1 L | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
2,2,4-trimethylpentane | |
10907 | |
114.23 | |
Liquid |
°C to 107°C | |
0.692 | |
-7°C (19°F) | |
>99.7% | |
C8H18 | |
MFCD00008943 | |
1696876 | |
Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,acetone,heptane and oils. Immiscible with water. | |
CC(C)CC(C)(C)C | |
114.232 | |
CHEBI:62805 | |
HPLC | |
2, 2, 4-Trimethylpentane |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Hazard Class: 3, Packing Group: II
EINECSNumber : 208-759-1
RTECSNumber : SA3320000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only